Phthalic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:88-99-3
:
Formula:C₈H₆O₄
MDL Number:MFCD00002467
:166.13 g/mol
Merck Index:12,07527
Boiling Pt:315…317 °C (1013 hPa)
Melting Pt:79 °C
Density:1.576 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:201-873-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:5.74 g/l (20 °C)
- Melting Point:191 °C (closed capillary tube)
- Molar Mass:166.13 g/mol
- Bulk Density:960 kg/m3
- Boiling Point:289 °C (1013 hPa) (decomposition)
- Vapor Pressure:7.8 hPa (191 °C)
- Flash Point:168 °C
- Density:1.59 g/cm3 (20 °C)
- pH:2 (5 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_822298
SDS
ProductPage_822298 ProductPage_822298
Product specification
Frequently Bought Together