Order Entry
Austria
ContactUsLinkComponent
 
Health
Irritant
Bis(2-ethylhexyl)phthalate, Supelco®
Catalog # 36735-1G
Supplier:  Merck
CAS Number:  
undefined
Health
Irritant
Bis(2-ethylhexyl)phthalate, Supelco®
Catalog # 36735-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :390.56 g/mol
Merck Index:13,02884
Boiling Pt:386.9 °C (1013 hPa)
Melting Pt:–50 °C
Density:0.98 g/cm³ (20 °C)
Flash Pt:195 °C
EINECS:204-211-0
UN:3082
ADR:9,III
 :M-Clarity™ quality level = MQ100
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:117-81-7
Formula:C₂₄H₃₈O₄
MDL Number:MFCD00009493