

Bis(2-ethylhexyl)phthalate
Catalog # EHERC16173000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
- Seq:3035
Specifications
About this item
:
:390.56 g/mol
Merck Index:13,02884
Boiling Pt:386.9 °C (1013 hPa)
Melting Pt:–50 °C
Density:0.98 g/cm³ (20 °C)
Flash Pt:195 °C
Storage Temperature:Ambient
EINECS:204-211-0
UN:3082
ADR:9,III
:GHS08
:GHS07
CAS Number:117-81-7
Formula:C₂₄H₃₈O₄
MDL Number:MFCD00009493