
Propyl-4-hydroxybenzoate sodium salt, Sigma-Aldrich®
Catalog # SIAL95038-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:202.19 g/mol
:M-Clarity™ quality level = MQ200
EINECS:252-488-1
:GHS05
Storage Temperature:Ambient
CAS Number:35285-69-9
Formula:C₁₀H₁₁NaO₃
MDL Number:MFCD00016477