Methyl-4-hydroxybenzoate, Sigma-Aldrich®
: Merck
:99-76-3
:
:HOC₆H₄CO₂CH₃
:MFCD00002352
:152.15 g/mol
:13,06129
:270…280 °C (1013 hPa)
:125…128 °C
:1.1208 g/cm³ (137 °C)
:M-Clarity™ quality level = MQ300
:202-785-7
:GHS07
:Ambient
Frequently Bought Together