Order Entry
Austria
ContactUsLinkComponent
Methyl-4-hydroxybenzoate ≥99%, TECHNICAL
Irritant
Methyl-4-hydroxybenzoate ≥99%, TECHNICAL
Catalog # 25605.293
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Methyl-4-hydroxybenzoate ≥99%, TECHNICAL
Catalog # 25605.293
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 99 %

Specification Test Results

About this item

 :
 :152.15 g/mol
Merck Index:13,06129
Boiling Pt:270…280 °C (1013 hPa)
Melting Pt:125…128 °C
Density:1.1208 g/cm³ (137 °C)
Storage Temperature:Ambient
EINECS:202-785-7
 :GHS07
CAS Number:99-76-3
Formula:HOC₆H₄CO₂CH₃
MDL Number:MFCD00002352