
Propyl benzoate, Sigma-Aldrich®
Catalog # SIAL307009-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
Boiling Pt:231 °C (1013 hPa)
Melting Pt:–52 °C
Density:1.02 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:>100 °C
:164.2 g/mol
Storage Temperature:Ambient
EINECS:219-020-8
:GHS07
CAS Number:2315-68-6
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00009370