Order Entry
Austria
ContactUsLinkComponent
Benzene-1,3,5-tricarbonyl trichloride for synthesis, Sigma-Aldrich®
Corrosive
Benzene-1,3,5-tricarbonyl trichloride for synthesis, Sigma-Aldrich®
Catalog # 8.41731.0025
Supplier:  Merck
CAS Number:  
Corrosive
Benzene-1,3,5-tricarbonyl trichloride for synthesis, Sigma-Aldrich®
Catalog # 8.41731.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Melting Point:
    34.5 - 36 °C
  • Molar Mass:
    265.47 g/mol
  • Boiling Point:
    180 °C (21 hPa)
  • Flash Point:
    112 °C

Specification Test Results

About this item

 :
 :265.48 g/mol
Boiling Pt:312 °C (1013 hPa)
Melting Pt:34…36 °C
Storage Temperature:Ambient
ADR:8,II
 :M-Clarity™ quality level = MQ200
EINECS:224-594-8
UN:3261
 :GHS05
CAS Number:4422-95-1
Formula:C₉H₃Cl₃O₃
MDL Number:MFCD00000679