
Benzene-1,3,5-tricarbonyl trichloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4422-95-1
:
Formula:C₉H₃Cl₃O₃
MDL Number:MFCD00000679
:265.48 g/mol
Boiling Pt:180 °C (16 mmHg)
Melting Pt:34.5…36 °C
Flash Pt:>112 °C
Storage Temperature:Ambient
EINECS:224-594-8
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to yellow Solid or liquid
- Infrared spectrum:Conforms
- Melting point:30 to 35 °C
- Titration Argentometric:97.5 to 102.5 % (ex Cl)
Specification Test Results
Frequently Bought Together