4-Chloro-2-iodoaniline, Sigma-Aldrich®
Catalog # SIAL516139-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:253.47 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:263-839-3
UN:2811
CAS Number:63069-48-7
Formula:C₆H₅ClIN
MDL Number:MFCD01863737