Order Entry
Austria
ContactUsLinkComponent
Affinity chromatography media, MabSelect™ PrismA
Affinity chromatography media, MabSelect™ PrismA
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography media, MabSelect™ PrismA
Supplier:  Cytiva

Specifications

  • pH range:
    3 – 12
  • Matrix:
    Rigid, highly cross-linked agarose
  • Ligand:
    Alkaline-stabilised protein A-derived (E.coli)
  • Storage temperature:
    2 - 8 °C
  • Binding capacity:
    ~ 40 mg polyclonal IgG/ml resin, 2 min residence time
    ~ 80 mg polyclonal IgG/ml resin, 6 min residence time
  • Ligand coupling method:
    Epoxy

Specifications

Related Information

Frequently Bought Together