Order Entry
Austria
ContactUsLinkComponent
2,3-Dichlorobenzaldehyde, Sigma-Aldrich®
2,3-Dichlorobenzaldehyde, Sigma-Aldrich®
Catalog # SIAL254835-25G
Supplier:  Merck
CAS Number:  
2,3-Dichlorobenzaldehyde, Sigma-Aldrich®
Catalog # SIAL254835-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

 :
 :175.01 g/mol
Boiling Pt:243
EINECS:228-711-3
 :GHS07
Melting Pt:64
Storage Temperature:Ambient
CAS Number:6334-18-5
Formula:C₇H₄Cl₂O
MDL Number:MFCD00010127
Density:1.33 g/cm³ (100 °C)
 :M-Clarity™ quality level = MQ200