Order Entry
Austria
Orders LinkContactUsLinkComponent
Vanillin | Avantor

Vanillin

Irritant
Marketsource
Anbieter:   Roth Carl
Vanillin
Anbieter:  Roth Carl
Marketsource
Certificates
Vanillin

2 Options Available Below

Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,09998
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:∼82 °C
  • Density:1.056 g/cm³ (20 °C)
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Product Family Options

Product Information

  • Pack type
    Pk
    Verfügbarkeit
    Preis
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 22,80 €
    22,80 €
    1 * 100 g
       22,80 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 120,00 €
    120,00 €
    1 * 1 kg
       120,00 €

    Specifications:
    Marketsource
    Marketsource