Order Entry
Austria
Orders LinkContactUsLinkComponent
Vanillin

Vanillin

Glass bottle for solids, 1 g
Catalog # ROTH5108.1
Irritant
Marketsource
Supplier:   Roth Carl
Vanillin
Catalog # ROTH5108.1
Supplier:  Roth Carl
Marketsource
CAS Number:  121-33-5
Vanillin
Vanillin

Glass bottle for solids, 1 g

CAS Number:  121-33-5
Price
109,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,09998
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:∼82 °C
  • Density:1.056 g/cm³ (20 °C)
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.