Order Entry
Austria
ContactUsLinkComponent
Indoline
Indoline
Catalog # MOLEM41324821
Supplier:  Molekula
CAS Number:  
Indoline
Catalog # MOLEM41324821
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2,3-Dihydroindol
 :
 :119.17 g/mol
Boiling Pt:220
Melting Pt:–21 °C
Density:1.0676 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-816-8
CAS Number:496-15-1
Formula:C₈H₉N
MDL Number:MFCD00005705