2,2,4-Trimethylpentane ≥99.8% (GC, on anhydrous basis), PESTINORM® Supra Trace
:540-84-1
:
:(CH₃)₂CHCH₂C(CH₃)₃
:MFCD00008943
:114.23 g/mol
:13,05212
:99.2 °C (1013 hPa)
:–107 °C
:0.692 g/cm³ (20 °C)
:Ambient
:208-759-1
:1262
:3,II
:Filtered through a 0.2 µm filter, packaged under inert gas.
:–12 °C
:GHS02
:GHS08
:GHS07
:GHS09
- Appearance:Clear colorless liquid
- Assay (GC, on anhydrous basis):min. 99.8 %
- Acidity (as Acetic acid):max. 0.001 %
- Color (APHA, Pt-Co):max. 5
- Residue after evaporation:max. 0.0001 % w/w
- Water (KF):max. 0.01 % w/w
- GC/ECD Dioxins, Furans & PCB's:max. 5 pg/ml
- GC/ECD any Pesticide (as Lindane):max. 5 pg/ml
- GC/NPD any Pesticide (as Parathion):max. 5 pg/ml
- GC/ECD 1,2,4-TCB to daca-PCB (as Lindane):max. 5 pg/ml
- GC/ECD DCM to 1,2,4-TCB (as TCM):max. 2 ng/ml
- GC/FID C10 to C40 (as n-Decane):max. 2 ng/ml
- GC/MSD C10 to C40 (as Decane; 30-600 amu):max. 2 ng/ml
Frequently Bought Together