Order Entry
Austria
ContactUsLinkComponent
Chloroform ≥99.9% (GC, on anhydrous basis) stabilised, PESTINORM® Supra Trace
Health
Toxic
Chloroform ≥99.9% (GC, on anhydrous basis) stabilised, PESTINORM® Supra Trace
Supplier:  VWR Chemicals
Certificates
undefined
Health
Toxic
Chloroform ≥99.9% (GC, on anhydrous basis) stabilised, PESTINORM® Supra Trace
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Clear colorless liquid
  • Color (APHA, Pt-Co):
    max. 10
  • Assay (GC, on anhydrous basis):
    min. 99.9 %
  • Acidity (as HCl):
    max. 0.0005 %
  • Residue after evaporation:
    max. 0.0003 % w/w
  • Stabilizer (Amylene):
    0.003-0.006 %w/w
  • Water (KF):
    max. 0.01 %w/w
  • GC/ECD Dioxins, Furans & PCB's:
    max. 5 pg/ml
  • GC/ECD any Pesticide (as Lindane):
    max. 5 pg/ml
  • GC/NPD any Pesticide (as Parathion):
    max. 10 pg/ml
  • GC/ECD 1,2,4-TCB to daca-PCB (as Lindane):
    max. 5 pg/ml
  • GC/FID C10 to C40 (as n-Decane):
    max. 3 ng/ml
  • GC/MSD C10 to C40 (as n-Decane; 30-600amu):
    max. 3 ng/ml

Specification Test Results

Frequently Bought Together