Cycloheximide
Supplier: PanReac AppliChem
Springe zu:
Über diesen Artikel
Synonyme: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
- CAS Nummer:66-81-9
- Formel:C₁₅H₂₃NO₄
- MDL-Nummer:MFCD00082346
- Molekulargewicht:281.35 g/mol
- Lagertemperatur:Ambient
- EINECS:200-636-0
- UN:2811
- ADR:6.1,I
3 Optionen unten verfügbar
Product Details & Documents
Synonyme: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
- CAS Nummer:66-81-9
- Formel:C₁₅H₂₃NO₄
- MDL-Nummer:MFCD00082346
- Molekulargewicht:281.35 g/mol
- Lagertemperatur:Ambient
- EINECS:200-636-0
- UN:2811
- ADR:6.1,I
Documents
Product Family Options
Produktinformationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



