

N-Methyl-2-pyrrolidone (NMP), Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:872-50-4
:
Formula:C₅H₉NO
MDL Number:MFCD00003193
:99.13 g/mol
Merck Index:13,06140
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:91 °C
Storage Temperature:Ambient
EINECS:212-828-1
:GHS08
:GHS07
Frequently Bought Together