Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Schiff reagent
Schiff reagent intense, Sigma-Aldrich®
Schiff reagent intense, Sigma-Aldrich®
Schiff reagent intense, Sigma-Aldrich®
1.02572.1000
:
Merck
:
Schiff reagent intense, Sigma-Aldrich®
1.02572.1000
:
Merck
:
Pk:
1 L
Seq:
5037
Pk Info:
Preparation Capacity 5000 - 6000 slides
:
:
1.014 g/cm3 (20 °C)
:
GHS07
:
MFCD00081985