Order Entry
Austria
ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP) for the electronics industry
N-Methyl-2-pyrrolidone (NMP) for the electronics industry
Catalog # 52010265.
CAS Number:  
undefined
N-Methyl-2-pyrrolidone (NMP) for the electronics industry
Catalog # 52010265.
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    210 kg
  • Seq:
    4684

Specifications

About this item

 :
 :99.13 g/mol
Merck Index:13,06140
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
Flash Pt:91 °C
EINECS:212-828-1
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:872-50-4
Formula:C₅H₉NO
MDL Number:MFCD00003193