Order Entry
Austria
ContactUsLinkComponent
 
N-Methyl-2-pyrrolidone (NMP) 99.5% for peptide synthesis
 
undefined
N-Methyl-2-pyrrolidone (NMP) 99.5% for peptide synthesis

 

  • Appearance:
    Clear liquid
  • Water:
    ≤0.1 % (K.F.)
  • GC:
    ≥99.4 %
  • Color scale:
    ≤10 APHA
  • Free amine:
    ≤0.01 %

 

 

Frequently Bought Together