Succinimide, Sigma-Aldrich®
SIALS9381-500G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
:
:99.09 g/mol
:12,09040
:285 °C (1013 hPa)
:121…124 °C
:1.418 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:Ambient
:204-635-6
:123-56-8
:C₄H₅NO₂
:MFCD00005495