Order Entry
Austria
ContactUsLinkComponent
Succinimide for synthesis, Sigma-Aldrich®
Succinimide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Succinimide for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    330 g/l (20 °C)
  • Melting Point:
    121 - 124 °C
  • Molar Mass:
    99.08 g/mol
  • Vapor Pressure:
     - 1 hPa (50 °C)
  • Flash Point:
    201 °C
  • Density:
    1.4 g/cm3 (20 °C)
  • pH:
    4 - 6 (200 g/l, H2O)
  • Hill Formula:
    C4H5NO2

Specification Test Results

Related Information

Frequently Bought Together