4-Octyne, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Dipropylacetylene
CAS Number:1942-45-6
:
Formula:C₈H₁₄
MDL Number:MFCD00009471
:110.2 g/mol
Boiling Pt:132 °C (1013 hPa)
Melting Pt:–103 °C
Density:0.7509 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:217-730-2
ADR:3,II
:M-Clarity™ quality level = MQ100
UN:1993
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together