Caffeic acid
Glass bottle for solids, 100 mg
Catalog # ROTH5574.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
- CAS Number:331-39-5
- Formula:C₉H₈O₄
- MDL Number:MFCD00004392
- Molecular Weight:180.16 g/mol
- Merck Index:12,01673
- Boiling Pt:366 °C (1013 hPa)
- Melting Pt:234…237 °C
- EINECS:206-361-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



