Order Entry
Austria
Orders LinkContactUsLinkComponent
Caffeic acid

Caffeic acid

Glass bottle for solids, 25 g
Catalog # MOLE77345158-25G
Marketsource
Supplier:   Molekula
Caffeic acid
Catalog # MOLE77345158-25G
Supplier:  Molekula
Marketsource
CAS Number:  331-39-5
Caffeic acid

Glass bottle for solids, 25 g

CAS Number:  331-39-5
Price
92,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
  • CAS Number:331-39-5
  • Formula:C₉H₈O₄
  • MDL Number:MFCD00004392
  • Molecular Weight:180.16 g/mol
  • Merck Index:12,01673
  • Boiling Pt:366 °C (1013 hPa)
  • Melting Pt:234…237 °C
  • EINECS:206-361-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.