Caffeic acid for synthesis, Sigma-Aldrich®
Catalog # 8.22029.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Hill Formula:C₉H₈O₄
- Chemical formula:(OH)₂C₆H₃CHCHCOOH
- Molar Mass:180.15 g/mol
- Melting Point:234 - 237 °C (decomposition)
- Solubility:(20 °C) slightly soluble
- Storage class:10 - 13 Other liquids and solids
- WGK:WGK 3 highly water endangering
- R Phrase:R 40
- S Phrase:S 36/37
Specification Test Results
About this item
:
:180.16 g/mol
Merck Index:12,01673
Boiling Pt:366 °C (1013 hPa)
Melting Pt:234…237 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:206-361-2
:GHS08
CAS Number:331-39-5
Formula:C₉H₈O₄
MDL Number:MFCD00004392