Order Entry
Austria
ContactUsLinkComponent
Caffeic acid, Millipore®
Caffeic acid, Millipore®
Catalog # 205546-500
CAS Number:  
Caffeic acid, Millipore®
Catalog # 205546-500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :180.16 g/mol
Merck Index:12,01673
Boiling Pt:366 °C (1013 hPa)
Melting Pt:234…237 °C
EINECS:206-361-2
CAS Number:331-39-5
Formula:C₉H₈O₄
MDL Number:MFCD00004392