2,3-Dimethyl-2-butene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Tetramethylethylene
CAS Number:563-79-1
:
Formula:C₆H₁₂
MDL Number:MFCD00008897
:84.16 g/mol
Boiling Pt:73 °C (1013 hPa)
Melting Pt:–75 °C
Density:0.7188 g/cm³ (20 °C)
Flash Pt:–16 °C
Storage Temperature:Refrigerator
UN:3295
ADR:3,II
:M-Clarity™ quality level = MQ100
EINECS:209-263-8
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together