2-Methyl-3-buten-2-ol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 3-Hydroxy-3-methyl-1-butene
CAS Number:115-18-4
:
Formula:C₅H₁₀O
MDL Number:MFCD00004470
Boiling Pt:99 °C (1013 hPa)
Melting Pt:–15 °C
Density:0.824 g/cm³ (20 °C)
Flash Pt:11 °C
EINECS:204-068-4
UN:1987
ADR:3,II
:M-Clarity™ quality level = MQ200
:86.13 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-28 °C
- Molar Mass:86.13 g/mol
- Boiling Point:98 - 99 °C (1013 hPa)
- Vapor Pressure:137 hPa (50 °C)
- Flash Point:11 °C
- Explosion Limit:1.5 - 9.4 %(V)
- Density:0.824 g/cm3 (20 °C)
- pH:9 - 10 (100 g/l, H2O, 20 °C)
- Ignition Point:380 °C
Specification Test Results
Frequently Bought Together