Order Entry
Austria
Orders LinkContactUsLinkComponent
Fumaric acid

Fumaric acid

Glass bottle for solids, 1 kg
Catalog # MOLE15604498-1KG
Irritant
Marketsource
Supplier:   Molekula
Fumaric acid
Catalog # MOLE15604498-1KG
Supplier:  Molekula
Marketsource
CAS Number:  110-17-8
Fumaric acid

Glass bottle for solids, 1 kg

CAS Number:  110-17-8
Price
115,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  trans-1,2-Ethylentricarboxylic acid
  • CAS Number:110-17-8
  • Formula:C₄H₄O₄
  • MDL Number:MFCD00002700
  • Molecular Weight:116.07 g/mol
  • Merck Index:13,04308
  • Boiling Pt:356 °C (1013 hPa)
  • Melting Pt:287 °C
  • Density:1.625 g/cm³ (20 °C)
  • Flash Pt:273 °C
  • EINECS:203-743-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications