
Fumaric acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-17-8
:
Formula:C₄H₄O₄
MDL Number:MFCD00002700
:116.07 g/mol
Merck Index:13,04308
Boiling Pt:356 °C (1013 hPa)
Melting Pt:287 °C
Density:1.625 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:273 °C
EINECS:203-743-0
:GHS07
Specification Test Results
- Solubility:4.9 g/l (20 °C)
- Melting Point:287 °C
- Molar Mass:116.07 g/mol
- Boiling Point:290 °C (1013 hPa) (sublimed)
- Vapor Pressure:- 0.001 hPa (20 °C)
- Flash Point:230 °C
- Density:1.64 g/cm3 (20 °C)
- pH:2.1 (4.9 g/l, H2O, 20 °C)
- Ignition Point:375 °C
Specification Test Results
Frequently Bought Together