Fumaric acid ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 110-17-8 Molecular Formula: HO2CCH=CHCO2H Formula Weight: 116.07 Flash Point: 273°(523°F) Odour: Odourless pH: 2.1 Melting Point: ca 299° subl. Beilstein Reference: 605763 Merck Reference: 14,4287 MDL No.: MFCD00002700
Synonyms: trans-1,2-Ethylentricarboxylic acid
- CAS Number:110-17-8
- Formula:C₄H₄O₄
- MDL Number:MFCD00002700
- Molecular Weight:116.07 g/mol
- Merck Index:14,4287
- Boiling Pt:356 °C (1013 hPa)
- Melting Pt:∼ 299 °C (subl.)
- Density:1.625
- Flash Pt:273 °C (523 °F)
- Storage Temperature:Ambient
- EINECS:203-743-0
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 110-17-8 Molecular Formula: HO2CCH=CHCO2H Formula Weight: 116.07 Flash Point: 273°(523°F) Odour: Odourless pH: 2.1 Melting Point: ca 299° subl. Beilstein Reference: 605763 Merck Reference: 14,4287 MDL No.: MFCD00002700
Synonyms: trans-1,2-Ethylentricarboxylic acid
- CAS Number:110-17-8
- Formula:C₄H₄O₄
- MDL Number:MFCD00002700
- Molecular Weight:116.07 g/mol
- Merck Index:14,4287
- Boiling Pt:356 °C (1013 hPa)
- Melting Pt:∼ 299 °C (subl.)
- Density:1.625
- Flash Pt:273 °C (523 °F)
- Storage Temperature:Ambient
- EINECS:203-743-0
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



