Order Entry
Austria
ContactUsLinkComponent
Maleic acid
Maleic acid
Catalog # ROTHK304.1
Supplier:  Roth Carl
CAS Number:  
undefined
Maleic acid
Catalog # ROTHK304.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

About this item

 :
 :116.07 g/mol
Merck Index:13,05726
Boiling Pt:256 °C (1013 hPa)
Melting Pt:130…131 °C
Density:1.59 g/cm³ (20 °C)
Flash Pt:127 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-742-5
UN:3261
ADR:8,III
 :GHS07
CAS Number:110-16-7
Formula:C₄H₄O₄
MDL Number:MFCD00063177