Order Entry
Austria
ContactUsLinkComponent
Melting point standard, phenyl salicylate
Melting point standard, phenyl salicylate
Catalog # 44770-5G
Supplier:  Merck
CAS Number:  
undefined
Melting point standard, phenyl salicylate
Catalog # 44770-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 g
  • Description:
    Phenyl salicylate
  • Melting pt.:
    41 – 43 °C (lit.)

Specifications

About this item

Mettler-Toledo calibration substance ME 30034252, traceable to primary standards.

  • Boiling point: 172 to 173 °C/12 mm Hg (lit.)
  • Format: neat
  • Limited shelf life

Synonym: Phenyl salicylate, salol