Triethylammonium bicarbonate buffers, for ion-exchange chromatography, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Buffer for use in ion-exchange chromatography and electrophoresis.
- Density: 1,02 g/ml at 25 °C (lit.)
- pH: 8,5±0,1
Triethylammonium bicarbonate (TEAB) is a buffer, composed of a combination of triethylamine and carbon dioxide, the latter occuring in solution as bicarbonate. The volatility of TEAB facilitates sample recovery after chromatographic analysis and makes TEAB a buffer of interest for mass spectrometric analysis of biomolecules.
Synonyms: TEAB, triethylammonium hydrogen, carbonate buffer.
Caution: Storage temperature: 2 to 8 °C
Specifications
- M-Clarity quality level:MQ200
Specifications
Frequently Bought Together