
(2-Methoxyphenyl)acetic acid for synthesis, Sigma-Aldrich®
8.18671.0025
: Merck
:
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
- Solubility:9.2 g/l (20 °C)
- Melting Point:120 - 123 °C
- Molar Mass:166.18 g/mol
:
:332 °C (1013 hPa)
:120…123 °C
:M-Clarity™ quality level = MQ200
:166.18 g/mol
:Ambient
:202-231-4
:GHS05
:93-25-4
:(C₆H₄OCH₃)CH₂COOH
:MFCD00004321