Order Entry
Austria
ContactUsLinkComponent
Phenylacetic acid for synthesis, Sigma-Aldrich®
Irritant
Phenylacetic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Phenylacetic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1.58 g/l (20 °C)
  • Melting Point:
    74 - 77 °C
  • Molar Mass:
    136.15 g/mol
  • Boiling Point:
    265 °C (1013 hPa)
  • Vapor Pressure:
    1.33 hPa (97 °C)
  • Flash Point:
    132 °C
  • Density:
    1.091 g/cm3 (97 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together