Ethyl phenylacetate, Sigma-Aldrich®
Catalog # SIAL108049-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:164.2 g/mol
Merck Index:13,03872
Boiling Pt:226 °C (1013 hPa)
Melting Pt:–29 °C
Density:1.03 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:202-993-8
CAS Number:101-97-3
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00009178