Order Entry
Austria
ContactUsLinkComponent
Ethyl phenylacetate
Ethyl phenylacetate
Catalog # ROTH4434.1
Supplier:  Roth Carl
CAS Number:  
Ethyl phenylacetate
Catalog # ROTH4434.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :164.2 g/mol
Merck Index:13,03872
Boiling Pt:226 °C (1013 hPa)
Melting Pt:–29 °C
Density:1.03 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-993-8
CAS Number:101-97-3
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00009178