
Indol-3-acetic acid 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:87-51-4
:
Formula:C₁₀H₉NO₂
MDL Number:MFCD00005636
:175.19 g/mol
Merck Index:13,04986
Boiling Pt:120 - 125 °C (0,05 -rr)
Melting Pt:167 °C
Flash Pt:171 °C
EINECS:201-748-2
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to beige to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:165 to 169 °C
- HPLC:≥99.0 %
- Elemental Analysis:3.72 to 3.99 % N
Specification Test Results
Frequently Bought Together