Order Entry
Austria
ContactUsLinkComponent
2,2,4-Trimethylpentane ≥99%, Reagent Grade
Irritant
Health
Flammable
Environment
2,2,4-Trimethylpentane ≥99%, Reagent Grade
Catalog # 258776-2.5L
CAS Number:  
undefined
Irritant
Health
Flammable
Environment
2,2,4-Trimethylpentane ≥99%, Reagent Grade
Catalog # 258776-2.5L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Autoignition Temperature:
    745 °F
  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.0 %
  • Vapor Density:
    3.9 (vs air)
  • Explosion Limit:
    6 %
  • Vapor Pressure:
    41 mmHg ( 21 °C)
  • Refractive Index:
    n20/D 1.391 (lit.)

Specification Test Results

About this item

 :
 :114.23 g/mol
Merck Index:13,05212
Boiling Pt:99 °C (1.013 hPa)
Melting Pt:-107 °C
Melting Pt:–107 °C
Density:0.69 g/cm³ (20 °C)
Flash Pt:-12 °C
Flash Pt:–12 °C
Storage Temperature:Ambient
EINECS:208-759-1
UN:1262
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
 :GHS09
CAS Number:540-84-1
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943