Order Entry
Austria
ContactUsLinkComponent
2,2,4-Trimethylpentane for synthesis, Sigma-Aldrich®
2,2,4-Trimethylpentane for synthesis, Sigma-Aldrich®
Catalog # 8.21627.1000
Supplier:  Merck
CAS Number:  
undefined
2,2,4-Trimethylpentane for synthesis, Sigma-Aldrich®
Catalog # 8.21627.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    0.56 mg/l (25 °C)
  • Melting Point:
    -107 °C
  • Molar Mass:
    114.23 g/mol
  • Boiling Point:
    99 °C (1013 hPa)
  • Vapor Pressure:
    51 hPa (20 °C)
  • Flash Point:
    -14 °C
  • Explosion Limit:
    1 - 5.5 %(V)
  • Density:
    0.69 g/cm3 (20 °C)
  • pH:
    (H2O) neutral
  • Ignition Point:
    410 °C
  • Saturation concentration:
    239 g/m3 (20 °C)
  • Evaporation number:
    2.3
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

About this item

 :
 :114.23 g/mol
Merck Index:13,05212
Boiling Pt:99.2 °C (1013 hPa)
Melting Pt:–107 °C
Density:0.692 g/cm³ (20 °C)
Flash Pt:–12 °C
Storage Temperature:Ambient
EINECS:208-759-1
UN:1262
ADR:3,II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:540-84-1
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943