Order Entry
Austria
ContactUsLinkComponent
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier:  Merck
Certificates
undefined
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Solubility:
    0.56 mg/l (25 °C)
  • Melting Point:
    -107 °C
  • Molar Mass:
    114.23 g/mol
  • Boiling Point:
    99 °C (1013 hPa)
  • Vapor Pressure:
    51 hPa (20 °C)
  • Flash Point:
    -14 °C
  • Explosion Limit:
    1 - 5.5 %(V)
  • Density:
    0.69 g/cm3 (20 °C)
  • pH:
    (H2O) neutral
  • Ignition Point:
    410 °C
  • Saturation concentration:
    239 g/m3 (20 °C)
  • Evaporation number:
    2.3
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

Related Information

Frequently Bought Together