ContactUsLinkComponent
Stopcocks, three-way, Supelco®
Stopcocks, three-way, Supelco®
Catalog # S7521-10EA
Supplier:  Merck
undefined
Stopcocks, three-way, Supelco®
Catalog # S7521-10EA
Supplier:  Merck
Specifications
  • Description:
    With female Luer-Lock inlet and male Luer-Lock outlet ports
  • Environmentally Preferable:
  • Pk:
    10
Specifications
About this item
Made of nylon.
  • Good chemical resistance
  • Good temperature resistance
Features three-way control with female luer-lock inlet and male luer-lock outlet ports.