Microtube racks, UniRack™ Jr
Supplier: Simport Scientific
Certificates
About this item
Made of polypropylene, coloured.
- Good chemical resistance
- Good temperature resistance
This smaller model of the UniRack can hold up to 25 PS or PP of 10 and 12 mm tubes, such as 10×75 mm or 12×75 mm sizes. Flip it over to hold up to 16 PCR tubes or microcentrifuge tubes from 0,2 to 0,5 ml. This rack will accommodate all types of screw cap microtubes from 0,5 to 2 ml and 1 to 5 ml cryogenic vials.
Delivery: Supplied without handles or anchor pins.
Frequently Bought Together