

2,2-Dimethyl-1-propanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-84-3
:
Formula:C₅H₁₂O
MDL Number:MFCD00004682
Merck Index:12,06546
Boiling Pt:113 °C (1013 hPa)
Density:0.81 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:4.1,II
:M-Clarity™ quality level = MQ200
:88.15 g/mol
EINECS:200-907-3
UN:1325
:GHS02
:GHS07
Frequently Bought Together