Guillard’s (F/2) marine water enrichment solution, Sigma-Aldrich®
Catalog # SIALG0154-500ML
Supplier: Merck
CAS Number:
Specifications
- Pk:500 mL
- Description:Guillard’s (F/2) marine water enrichment solution, 50X
Specifications
About this item
Guillard’s (F/2) marine water enrichment solution 50X, is a concentrate of major nutrients, trace metals and vitamins that can be diluted in filtered seawater to support reconstitution of Guillard’s (F/2) marine enrichment medium.
Guillard’s (F/2) marine enrichment medium is a widely used enriched seawater medium for growing costal marine algae (diatoms).
Reconstitution: Use at 20 ml/L of seawater.