Orders LinkContactUsLinkComponent
Filter funnels, micro
Filter funnels, micro
35 mm
Artikelnr. HACH2584335
Anbieter:   Hach
Filter funnels, micro
Artikelnr. HACH2584335
Anbieter:  Hach
Filter funnels, micro
Filter funnels, micro
35 mm
Preis
8,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
PP, translucent.
  • Good chemical resistance
  • Good temperature resistance
Ideal for micro work, filling burets, manometers, and small bottles.
Specifications
  • Int.Ø top:
    35 mm